Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gst-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306528.3 | NP_001293457.1 | 2 - 3 | 230 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |