Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dyb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306500.3 | NP_001293429.1 | 2 - 3 | 196 | 93 | |
NM_001383732.1 | 3 - 4 | 446 | 93 | |||
NM_058459.6 | NP_490860.1 | 3 - 4 | 453 | 93 | ||
GenBank mRNA | AJ131742.1 | - | 3 - 4 | 430 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |