Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mab-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306514.3 | NP_001293443.1 | 5 - 6 | 1456 | 106 | |
NM_001306515.3 | NP_001293444.1 | 4 - 5 | 1285 | 106 | ||
NM_058474.5 | NP_490875.1 | 6 - 7 | 2018 | 106 | ||
NR_165326.1 | 5 - 6 | 2703 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |