Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058476.5 | NP_490877.1 | 1 - 2 | 389 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |