Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mppa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383757.1 | 3 - 4 | 1044 | 54 | ||
NM_058487.7 | NP_490888.2 | 3 - 4 | 1040 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |