Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mppa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383757.1 | 4 - 5 | 1396 | 75 | ||
NM_058487.7 | NP_490888.2 | 4 - 5 | 1392 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Caenorhabditis elegans Probe spans exons |