Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arl-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037897.3 | NP_001032986.1 | 8 - 9 | 959 | 101 | |
NM_001383787.1 | 7 - 8 | 828 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |