Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306399.3 | NP_001293328.1 | 11 - 12 | 1706 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |