Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306398.2 | NP_001293327.1 | 14 - 15 | 2776 | 78 | |
NM_001306399.3 | NP_001293328.1 | 14 - 15 | 2767 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |