Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306398.2 | NP_001293327.1 | 9 - 10 | 1511 | 66 | |
NM_001306399.3 | NP_001293328.1 | 9 - 10 | 1511 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |