Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dct-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058543.4 | NP_490944.1 | 2 - 3 | 221 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |