Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nekl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306391.3 | NP_001293320.1 | 1 - 2 | 111 | 94 | |
NR_131355.1 | 1 - 2 | 111 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |