Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026642.5 | NP_001021813.1 | 5 - 6 | 1637 | 70 | |
NM_001306438.3 | NP_001293367.1 | 2 - 3 | 401 | 70 | ||
NM_058627.7 | NP_491028.2 | 4 - 5 | 636 | 70 | ||
NR_131370.1 | 2 - 3 | 435 | 70 | |||
GenBank mRNA | U63557.1 | - | 4 - 5 | 645 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Caenorhabditis elegans Probe spans exons |