Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: itx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001262315.4 | NP_001249244.2 | 23 - 24 | 4070 | 74 | |
NM_001262316.5 | NP_001249245.2 | 24 - 25 | 4014 | 74 | ||
NM_001306453.2 | NP_001293382.1 | 24 - 25 | 4014 | 74 | ||
NM_001306454.3 | NP_001293383.1 | 21 - 22 | 3494 | 74 | ||
NM_001306455.3 | NP_001293384.1 | 21 - 22 | 3494 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |