Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccb-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306451.2 | NP_001293380.1 | 4 - 5 | 397 | 88 | |
NM_001306452.1 | NP_001293381.1 | 4 - 5 | 397 | 88 | ||
NR_131378.1 | 4 - 5 | 580 | 88 | |||
NR_131379.1 | 4 - 5 | 440 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |