Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bam-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026638.4 | NP_001021809.1 | 5 - 6 | 1030 | 97 | |
GenBank mRNA | AY373340.1 | - | 5 - 6 | 996 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |