Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058683.4 | NP_491084.1 | 1 - 2 | 220 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |