Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mod-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058694.4 | NP_491095.3 | 10 - 11 | 1715 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |