Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025871.6 | NP_001021042.2 | 8 - 9 | 891 | 70 | |
NM_001025872.6 | NP_001021043.2 | 6 - 7 | 785 | 70 | ||
NM_001356801.3 | NP_001343721.1 | 7 | 873 | 70 | ||
GenBank mRNA | AY204173.1 | - | 7 - 8 | 882 | 70 | |
AY204174.1 | - | 6 - 7 | 807 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |