Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fog-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313540.3 | NP_001300469.1 | 3 - 4 | 820 | 67 | |
NM_001381585.1 | NP_001367957.1 | 8 - 9 | 1639 | 67 | ||
NM_001381586.2 | NP_001367956.1 | 8 - 9 | 1645 | 67 | ||
GenBank mRNA | AJ297846.1 | - | 8 - 9 | 1661 | 67 | |
AJ297847.1 | - | 8 - 9 | 1661 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |