Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cogc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058727.6 | NP_491128.4 | 6 - 7 | 912 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |