Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sep-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058759.6 | NP_491160.1 | 9 - 10 | 3081 | 59 | |
GenBank mRNA | AF399825.1 | - | 8 - 9 | 3058 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |