Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lpd-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313535.2 | NP_001300464.1 | 27 - 28 | 10718 | 70 | |
NM_001313536.3 | NP_001300465.1 | 27 - 28 | 10728 | 70 | ||
NM_001313537.3 | NP_001300466.1 | 27 - 28 | 10670 | 70 | ||
NM_001313538.2 | NP_001300467.1 | 27 - 28 | 10728 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |