Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mis-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129068.3 | NP_001122540.1 | 4 - 5 | 337 | 52 | |
NM_058783.8 | NP_491184.1 | 4 - 5 | 296 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |