Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cah-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058788.3 | NP_491189.1 | 5 - 6 | 973 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |