Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058846.5 | NP_491247.2 | 6 - 7 | 848 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |