Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inx-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026421.4 | NP_001021592.1 | 2 - 3 | 362 | 69 | |
NM_058914.4 | NP_491315.1 | 2 - 3 | 302 | 69 | ||
GenBank mRNA | AF303262.1 | - | 2 - 3 | 302 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |