Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: met-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058939.6 | NP_491340.2 | 4 - 5 | 728 | 86 | |
NM_182042.4 | NP_871842.2 | 4 - 5 | 699 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |