Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330925.3 | NP_001317773.1 | 1 - 2 | 1411 | 99 | |
NM_058962.6 | NP_491363.1 | 4 - 5 | 4220 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |