Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026429.3 | NP_001021600.1 | 11 - 12 | 2727 | 73 | |
NM_001262699.2 | NP_001249628.1 | 14 - 15 | 2621 | 73 | ||
NM_001372632.2 | NP_001360115.1 | 14 - 15 | 2613 | 73 | ||
NM_001382937.1 | 10 - 11 | 2518 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |