Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026429.3 | NP_001021600.1 | 2 - 3 | 1019 | 84 | |
NM_001262699.2 | NP_001249628.1 | 6 - 7 | 949 | 84 | ||
NM_001372632.2 | NP_001360115.1 | 6 - 7 | 935 | 84 | ||
NM_001382937.1 | 2 - 3 | 846 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |