Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059090.9 | NP_491491.2 | 6 - 7 | 1239 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |