Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dyf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059093.7 | NP_491494.2 | 12 - 13 | 1746 | 117 | |
NM_182063.7 | NP_871863.1 | 10 - 11 | 1527 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |