Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lfe-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025878.7 | NP_001021049.1 | 4 - 5 | 349 | 61 | |
NM_001262849.3 | NP_001249778.1 | 4 - 5 | 332 | 61 | ||
NM_001372686.1 | NP_001359911.1 | 3 - 4 | 271 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |