Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mat-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026543.7 | NP_001021714.1 | 5 - 6 | 849 | 73 | |
NM_001377581.3 | NP_001364721.1 | 4 - 5 | 766 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |