Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059132.5 | NP_491533.2 | 1 - 2 | 118 | 81 | |
GenBank mRNA | AF288374.1 | - | 1 - 2 | 116 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |