Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059132.5 | NP_491533.2 | 4 - 5 | 456 | 62 | |
GenBank mRNA | AF288374.1 | - | 4 - 5 | 454 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |