Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frm-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001262958.3 | NP_001249887.1 | 16 - 17 | 1720 | 82 | |
NM_001383090.1 | 15 - 16 | 1635 | 82 | |||
NM_059185.7 | NP_491586.1 | 14 - 15 | 1611 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |