Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: che-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059272.6 | NP_491673.1 | 6 - 7 | 1112 | 72 | |
GenBank mRNA | AJ277649.1 | - | 6 - 7 | 1020 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |