Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383147.1 | 5 - 6 | 635 | 155 | ||
GenBank mRNA | AB027412.1 | - | 5 - 6 | 639 | 155 | |
AF153275.1 | - | 5 - 6 | 575 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |