Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383147.1 | 7 - 8 | 827 | 57 | ||
GenBank mRNA | AB027412.1 | - | 7 - 8 | 831 | 57 | |
AF153275.1 | - | 7 - 8 | 767 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |