Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059285.4 | NP_491686.1 | 9 - 10 | 2277 | 129 | |
GenBank mRNA | AF116529.1 | - | 9 - 10 | 2299 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |