Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028198.5 | NP_001023369.1 | 4 - 5 | 503 | 62 | |
NM_001307204.3 | NP_001294133.1 | 1 - 2 | 83 | 62 | ||
NM_001307207.3 | NP_001294136.1 | 2 - 3 | 263 | 62 | ||
NM_001383139.1 | 1 - 2 | 333 | 62 | |||
NM_171396.7 | NP_741462.1 | 5 - 6 | 730 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |