Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028198.5 | NP_001023369.1 | 7 - 8 | 797 | 92 | |
NM_001307204.3 | NP_001294133.1 | 4 - 5 | 377 | 92 | ||
NM_001307205.2 | NP_001294134.1 | 8 - 9 | 1038 | 92 | ||
NM_001307206.3 | NP_001294135.1 | 7 - 8 | 791 | 92 | ||
NM_001307207.3 | NP_001294136.1 | 5 - 6 | 557 | 92 | ||
NM_001380356.1 | NP_001368431.1 | 4 - 5 | 371 | 92 | ||
NM_001383138.1 | 8 - 9 | 1125 | 92 | |||
NM_001383139.1 | 4 - 5 | 627 | 92 | |||
NM_171396.7 | NP_741462.1 | 8 - 9 | 1024 | 92 | ||
NM_171397.8 | NP_741463.1 | 5 - 6 | 551 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |