Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttll-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380378.1 | NP_001367737.1 | 8 - 9 | 1207 | 92 | |
NM_171405.7 | NP_741471.2 | 10 - 11 | 1504 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |