Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klp-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136382.3 | NP_001129854.3 | 3 - 4 | 294 | 78 | |
NM_001136383.3 | NP_001129855.3 | 1 - 2 | 99 | 78 | ||
NM_001383153.1 | 13 - 14 | 2347 | 78 |
Gene Symbol: F20C5.11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383152.1 | 13 - 14 | 2352 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |