Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028074.5 | NP_001023245.1 | 6 - 7 | 1093 | 78 | |
NM_001136387.4 | NP_001129859.1 | 4 - 5 | 880 | 78 | ||
NM_069315.6 | NP_501716.2 | 6 - 7 | 1046 | 78 | ||
NM_171412.4 | NP_741480.1 | 4 - 5 | 749 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |