Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pxf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028218.6 | NP_001023389.1 | 8 - 9 | 2800 | 148 | |
NM_001028219.3 | NP_001023390.1 | 8 - 9 | 2800 | 148 | ||
NM_001268536.3 | NP_001255465.1 | 4 - 5 | 2420 | 148 | ||
NM_001268537.3 | NP_001255466.1 | 3 - 4 | 2294 | 148 | ||
NM_001380435.1 | NP_001367298.1 | 4 - 5 | 2420 | 148 | ||
NM_001380436.1 | NP_001367299.1 | 3 - 4 | 2294 | 148 | ||
GenBank mRNA | AF170796.1 | - | 8 - 9 | 2789 | 148 | |
AF308447.1 | - | 8 - 9 | 2789 | 148 | ||
AF308448.1 | - | 8 - 9 | 2789 | 148 | ||
AF308449.1 | - | 5 - 6 | 2487 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |