Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mlk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313222.3 | NP_001300151.1 | 12 - 13 | 2219 | 74 | |
NM_171456.8 | NP_741536.1 | 11 - 12 | 1982 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |