Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mlk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313222.3 | NP_001300151.1 | 13 - 14 | 2272 | 69 | |
NM_171456.8 | NP_741536.1 | 12 - 13 | 2035 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |